Learn More
Selleck Chemical LLC PRN1371 2mg 1802929-43-6
SDP
Supplier:  Selleck Chemical LLC S85782MG
Encompass_Preferred
PRN1371 is an irreversible covalent FGFR1-4 kinase inhibitor, with IC50s of 0.6, 1.3, 4.1, 19.3 and 8.1 nM for FGFR1, 2, 3, 4 and CSF1R, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8159