Learn More
TARGETMOL CHEMICALS INC PRN1371 5MG
SDP
Catalog No. 502578847
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PRN1371 is a specific and potent FGFR1-4 and CSF1R inhibitor (IC50s 0.6/1.3/4.1/19.3/8.1 nM for FGFR1/2/3/4 and CSF1R). purity: 99%
Catalog No. 50-257-8847 Supplier TARGETMOL CHEMICALS INC Supplier No. TQ00155MG
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PRN1371 is a specific and potent FGFR1-4 and CSF1R inhibitor (IC50s 0.6/1.3/4.1/19.3/8.1 nM for FGFR1/2/3/4 and CSF1R). purity: 99%