Learn More
Cayman Chemical Pranlukast-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 2713172-43-9, 1 mg
SDP
Catalog No. 502937480
Encompass_Preferred
An internal standard for the quantification of pranlukast by GC- or LC-MS
Catalog No. 50-293-7480 Supplier Cayman Chemical Supplier No. 287091 MG
An internal standard for the quantification of pranlukast by GC- or LC-MS