Learn More
Cayman Chemical Pranoprofen, Purity: ≥98%, CAS: 52549-17-4, 100 mg
SDP
Catalog No. 502979791
Encompass_Preferred
An NSAID and COX-1 inhibitor (IC50 = 3.1 µM); decreases B. pertussis-induced PGE2 production in isolated rat peritoneal leukocytes (IC50 = 0.39 µM); reduces corneal epithelial damage in a mouse model of corneal alkali burns when applied ocularly
Catalog No. 50-297-9791 Supplier Cayman Chemical Supplier No. 17875100 MG
An NSAID and COX-1 inhibitor (IC50 = 3.1 µM); decreases B. pertussis-induced PGE2 production in isolated rat peritoneal leukocytes (IC50 = 0.39 µM); reduces corneal epithelial damage in a mouse model of corneal alkali burns when applied ocularly