Learn More
Cayman Chemical Pranoprofen-13C-d3, Purity: ≥99% deuterated forms (d1-d3), 1 mg
SDP
Catalog No. 502950742
Encompass_Preferred
An internal standard for the quantification of pranoprofen by GC- or LC-MS
Catalog No. 50-295-0742 Supplier Cayman Chemical Supplier No. 340091 MG
An internal standard for the quantification of pranoprofen by GC- or LC-MS