Learn More
Cayman Chemical Prnoprofen-13C-d3 500ug
SDP
Catalog No. 502947497
Encompass_Preferred
An internal standard for the quantification of pranoprofen by GC- or LC-MS
Catalog No. 50-294-7497 Supplier Cayman Chemical Supplier No. 34009500 UG
An internal standard for the quantification of pranoprofen by GC- or LC-MS