Learn More
Honeywell™ PRO 3003 Leather Station/Duty Boots
Catalog No. 19814339
Change view
Click to view available options
Size:
10.5D
10D
11.5D
11D
12D
8.5D
9.5D
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-814-339 10D
19-168-827 9.5D
19-168-829 11D
19-168-831 12D
19-168-830 11.5D
19-168-828 10.5D
19-168-826 8.5D
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19-814-339 Supplier Honeywell™ Supplier No. BT30031002
Add to Cart
Add to Cart
Extremely lightweight and comfortable
Its oblique toe, Cambrelle lining, and stitched-down sole coupled with Military AB leather create a boot that is the most water-resistant available in the fire service. The unique design makes this the perfect boot for station/duty use.
  • Water resistant leather for durability, breathability and comfort
  • Oblique-shaped steel toe for added protection and comfort
  • Reinforced stitched-down sole
  • Abrasion and bacteria resistant Cambrelle lining
  • Foam-filled comfort collar that will not chafe
  • Dual function closure system for enhanced ankle support and fit plus zipper for rapid donning and doffing
  • Thermoplastic backpart molded heel counter ensures superior comfort, fit and durability; does not lose shape if exposed to moisture
  • Full wraparound Goodyear™ welt adds support, reduces fatigue and ankle sprains/strains
  • Welted construction retains its shape over the life of the boot so you keep that great fit
  • Triple linear ridge shank for increased arch support and puncture resistant steel bottom plate
  • Vibram Technical Lug non-marking outsole, combined with special fire retardant NFPA compound, for improved traction and durability
  • Full wrap-around Goodyear welt adds support and reduces fatigue and ankle strains and sprains
  • Nonmarking Vibram™ lug sole
  • Thermoplastic backpart molded heel counter maintains shape in moist conditions
  • Triple linear ridge shank for increased arch support
  • Puncture-resistant steel bottom plate
  • 8 in. high
This boot does not comply with any NFPA standards
Order Info
D width shown; also available in E and EEE widths.
TRUSTED_SUSTAINABILITY
Specifications
Height (English) 8 in.
Liner Material Cambrelle
Material Leather
Size 10D