Learn More
Honeywell™ PRO 5006 Structural Firefighting Boots, D Width
Catalog No. 19167403
Change view
Click to view available options
Material:
Leather, Dupont Kevlar Aramid Fiber, CrossTech
Pro-V Liner
Size:
10.5D
10D
11.5D
11D
12.5D
12D
13D
14D
8.5D
8D
9.5D
9D
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Size
19-167-403 Pro-V Liner 8.5D
19-167-413 Pro-V Liner 14D
19-167-405 Pro-V Liner 9.5D
19-167-409 Leather, Dupont Kevlar Aramid Fiber, CrossTech 11.5D
19-167-412 Pro-V Liner 13D
19-167-410 Pro-V Liner 12D
19-167-408 Pro-V Liner 11D
19-167-402 Pro-V Liner 8D
19-167-406 Pro-V Liner 10D
19-167-407 Leather, Dupont Kevlar Aramid Fiber, CrossTech 10.5D
19-167-411 Leather, Dupont Kevlar Aramid Fiber, CrossTech 12.5D
19-167-404 Pro-V Liner 9D
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 19-167-403 Supplier Honeywell™ Supplier No. BT5006B0852
Add to Cart
Add to Cart
Superior combination of comfort and absorbency
Quilted to 7.5 oz Kevlar™ felt thermal barrier and laminated to fully encapsulating Crosstech™ Footwear Fabric bootie. Crosstech™ moisture barrier provides superior liquid penetration resistance to blood, body fluids and NFPA tested common chemicals.
  • Military AB leather construction insulates and adds abrasion-, puncture-, water- and chemical-resistance
  • With heavy duty 0.75 in. reinforced straps mounted to the exterior of boot and anchored with a 2 × 1.5 in. shield-shaped base
  • External texon board shinguard protects and guards sensitive shin area of legs
  • Patented PowerToe™ cover resists toe abrasion and provides unparalled wear over the lifetime of the boots
  • Oblique toe safety cap provides additional room in height and width of boot
  • Fire/Ice PowerSole™ angled lugs provide substantial edging and gripping capabilities
  • Molded thermoplastic heel counter locks foot into the boot and anchors it in place
  • Patented PowerHeel™ cover protects heel area from abusive doffing of boots
  • Corrison-resistant steel ladder shank and stainless steel, full length, toe-to-heel puncture resistant bottom plate
Order Info
D-widths shown in table below. Wide widths also available. Available through Fisher Safety's GSA Advantage!™ Schedule 84.
Compliance
Meet NFPA 1971, 2007 Edition. Berry Amendment compliant.
TRUSTED_SUSTAINABILITY
Specifications
Height (English) 14 in.
Material Pro-V Liner
Size 8.5D