Learn More
Medchemexpress LLC PRO-6E MET Degrader | 2353493-69-1 | 99.9% | 848.75 | 1 ML
SDP
Catalog No. 5000393976
Encompass_Preferred
PRO-6E is an oral active PROTAC based on a Cereblon ligand that induces the degradation of MET with a maximum degradation of 81.9% at 1 μM in MKN-45 cells. It inhibits tumor growth both in vivo and in vitro, and also induces cell apoptosis and cell arrest.
  • Oral active PROTAC
  • Induces degradation of MET
  • Inhibits tumor growth in vivo and in vitro
  • Induces cell apoptosis and cell arrest
Catalog No. 50-003-93976 Supplier Medchemexpress LLC Supplier No. HY16050610MM/1ML
PRO-6E is an oral active PROTAC based on a Cereblon ligand that induces the degradation of MET with a maximum degradation of 81.9% at 1 μM in MKN-45 cells. It inhibits tumor growth both in vivo and in vitro, and also induces cell apoptosis and cell arrest.
  • Oral active PROTAC
  • Induces degradation of MET
  • Inhibits tumor growth in vivo and in vitro
  • Induces cell apoptosis and cell arrest