Learn More
Medchemexpress LLC Pro-Ac and arginine buffer | 5 ML
SDP
Catalog No. 5000204017
Encompass_Preferred
100mM Pro-Ac, 20 mM Arg, pH 5.0 buffer is a ready-to-use aqueous buffer solution intended for laboratory research use. It contains 100 mM Pro-Ac (proline acetate) and 20 mM arginine and is supplied at pH 5.0. Not for human use.
Catalog No. 50-002-04017 Supplier Medchemexpress LLC Supplier No. HYY19775ML
100mM Pro-Ac, 20 mM Arg, pH 5.0 buffer is a ready-to-use aqueous buffer solution intended for laboratory research use. It contains 100 mM Pro-Ac (proline acetate) and 20 mM arginine and is supplied at pH 5.0. Not for human use.