Learn More
Medchemexpress LLC Pro-Ac buffer | 99.3% | 10 ML
SDP
Catalog No. 5000203889
Encompass_Preferred
100 mM Pro-Ac, pH 5.0 buffer is a biochemical reagent intended for use as a reconstitution buffer for antibody-drug conjugate (ADC) products and other research applications. Supplied as a liquid, it is colorless to light yellow and is shipped on dry ice for storage at -80°C.
  • Contains 100 mM Pro-Ac at pH 5.0 for acidic buffering
  • High reported purity suitable for research applications
  • Available in small-volume liquid formats for reconstitution workflows
  • Colorless to light yellow appearance for easy visual inspection
  • Shipped on dry ice and requires storage at -80°C to maintain stability
Catalog No. 50-002-03889 Supplier Medchemexpress LLC Supplier No. HYY1977A10ML
100 mM Pro-Ac, pH 5.0 buffer is a biochemical reagent intended for use as a reconstitution buffer for antibody-drug conjugate (ADC) products and other research applications. Supplied as a liquid, it is colorless to light yellow and is shipped on dry ice for storage at -80°C.
  • Contains 100 mM Pro-Ac at pH 5.0 for acidic buffering
  • High reported purity suitable for research applications
  • Available in small-volume liquid formats for reconstitution workflows
  • Colorless to light yellow appearance for easy visual inspection
  • Shipped on dry ice and requires storage at -80°C to maintain stability