Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pro-Ac buffer | 99.3% | 5 ML
SDP

Catalog No. 5000203890
Encompass_Preferred

100 mM Pro-Ac, pH 5.0 buffer is a ready-to-use liquid biochemical reagent formulated at 100 mM and adjusted to pH 5.0. It is intended as a reconstitution buffer for antibody-drug conjugates (ADCs) and related biochemical applications; supplied for research use only.

  • Concentration: 100 mM, pH 5.0.
  • Format: liquid; colorless to light yellow.
  • Purity: 99.29% (manufacturer reported).
  • Intended use: reconstitution buffer for ADC products; for research use only.
  • Available sizes: 5 mL, 10 mL, 50 mL, 100 mL; larger sizes available by quote.

Catalog No. 50-002-03890 Supplier Medchemexpress LLC Supplier No. HYY1977A5ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

100 mM Pro-Ac, pH 5.0 buffer is a ready-to-use liquid biochemical reagent formulated at 100 mM and adjusted to pH 5.0. It is intended as a reconstitution buffer for antibody-drug conjugates (ADCs) and related biochemical applications; supplied for research use only.

  • Concentration: 100 mM, pH 5.0.
  • Format: liquid; colorless to light yellow.
  • Purity: 99.29% (manufacturer reported).
  • Intended use: reconstitution buffer for ADC products; for research use only.
  • Available sizes: 5 mL, 10 mL, 50 mL, 100 mL; larger sizes available by quote.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.