Learn More
Medchemexpress LLC Pro-Ac buffer | 99.3% | 50 ML

Supplier: Medchemexpress LLC HYY1977A50ML
100 mM Pro-Ac, pH 5.0 buffer is an aqueous buffer solution prepared at 100 mM concentration and adjusted to pH 5.0 for use as a reconstitution buffer in research applications, including antibody-drug conjugate handling. It is supplied as a liquid and should be stored and handled according to the certificate of analysis and safety data sheet.
- Concentration: 100 mM aqueous buffer
- pH adjusted to 5.0 for acidic reconstitution conditions
- Appearance: colorless to light yellow liquid
- High purity suitable for research use
- Available in laboratory volumes, including 50 mL
- Store at -80°C as indicated on the certificate of analysis
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.