Learn More
CPC Scientific H-Ala-Arg-Leu-Asp-Val-Ala-Ala-Glu-Phe-Arg-Lys-Lys-Trp-Asn-Lys-Trp-Ala-Leu-Ser-Arg-NH2 (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific ADRM013A
Encompass
Prodepin, porcine.ONE-LETTER SEQUENCE: ARLDVAAEFRKKWNKWALSR-NH2MOLECULAR FORMULA: C112H178N36O26MOLECULAR WEIGHT:2444.9STORAGE CONDITIONS: -20 5CRESEARCH AREA: Cardiovascular
Catalog No. 50-210-8639