Learn More
CPC Scientific H-Ala-Arg-Leu-Asp-Thr-Ser-Ser-Gln-Phe-Arg-Lys-Lys-Trp-Asn-Lys-Trp-Ala-Leu-Ser-Arg-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific ADRM011B
Encompass
Prodepin, rat. Biological activity still unknown. High possibility that it is synthesized in vivo and holds some important physiological function.ONE-LETTER SEQUENCE: ARLDTSSQFRKKWNKWALSR-NH2MOLECULAR FORMULA: C111H177N37O28MOLECULAR WEIGHT:2477.9STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [167699-60-7], RESEARCH AREA: CardiovascularREFERENCES: J. Sakata et al., BBRC, 195, 921 (1993)
Catalog No. 50-210-8636