Learn More
Cell Signaling Technology Pro-Apoptosis Bcl-2 Family Antibody Sampler Kit, 1 Kit
SDP
Catalog No. 502729477
Encompass
The Pro-Apoptosis Bcl-2 Family Antibody Sampler Kit provides an economical means to examine several members of the Bcl-2 family and their activation status. The kit contains a comprehensive selection of primary antibodies and an HRP-linked secondary antibody, with enough material for two Western blot experiments per primary antibody.
  • Includes antibodies targeting pro-apoptotic proteins including Bad, Phospho-Bad (Ser75), Bax, Bik, Bid, Bim, Bak1, and PUMA.
  • Supplied with an anti-rabbit IgG, HRP-linked secondary antibody for convenient detection.
  • Validated for diverse research applications including Western blotting and immunoprecipitation.
  • Stored in a stabilized buffer with glycerol for optimal performance at −20°C.
Catalog No. 50-272-9477 Supplier Cell Signaling Technology Supplier No. 9942T
The Pro-Apoptosis Bcl-2 Family Antibody Sampler Kit provides an economical means to examine several members of the Bcl-2 family and their activation status. The kit contains a comprehensive selection of primary antibodies and an HRP-linked secondary antibody, with enough material for two Western blot experiments per primary antibody.
  • Includes antibodies targeting pro-apoptotic proteins including Bad, Phospho-Bad (Ser75), Bax, Bik, Bid, Bim, Bak1, and PUMA.
  • Supplied with an anti-rabbit IgG, HRP-linked secondary antibody for convenient detection.
  • Validated for diverse research applications including Western blotting and immunoprecipitation.
  • Stored in a stabilized buffer with glycerol for optimal performance at −20°C.