Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YMC America, Inc PRO C18 3UM 120A 10X2.1MM PK/5
SDP

Catalog No. 50157231
Encompass

AS12S03-01Q1GC: Pro C18, 3um, 120A, 10X2.1mm Cartridge, pk/5

Catalog No. 50-157-231 Supplier YMC America, Inc Supplier No. AS12S0301Q1GC
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AS12S03-01Q1GC: Pro C18, 3um, 120A, 10X2.1mm Cartridge, pk/5

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.