Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YMC America, Inc PRO C18 RS 5UM 80A 100X3MM
SDP

Catalog No. 50874628
Encompass_Preferred

Pro C18 Rs 5Um; 80A 100 X 3mm

Catalog No. 50-874-628 Supplier YMC America, Inc Supplier No. RS08S051003WT
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pro C18 Rs 5Um; 80A 100 X 3mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.