Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YMC America, Inc PRO C8 5UM 120A 100X2MM
SDP

Catalog No. 50874549
Encompass_Preferred

Pro C8 5Um; 120A; 100 X 2mm

Catalog No. 50-874-549 Supplier YMC America, Inc Supplier No. OS12S051002WT
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pro C8 5Um; 120A; 100 X 2mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.