Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YMC America, Inc PRO C8 5UM 120A 100X30MM ACTUS
SDP

Supplier:  YMC America, Inc OS12S051030WX

Encompass

Pro C8 5Um; 120A; 100 X 30mm Actus

Catalog No. 50-420-899


May include imposed supplier surcharges.
Only null left
Add to Cart
Request a Quote

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.