Learn More
Sigma Aldrich Fine Chemicals Biosciences ProClin 300, 2 L
Catalog No. NC0377211
Encompass
ProClin biocides are immediately microstatic upon contact with microbial organisms as a result of their ability to quickly penetrate cell membranes and inhibit specific enzymes crucial to cellular respiration. Multiple specific sites of ProClin toxicity prevent microorganisms from developing high levels of resistance, making ProClin highly efficient even over extended storage periods of your IVD components.
Catalog No. NC0377211 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 48915U
ProClin biocides are immediately microstatic upon contact with microbial organisms as a result of their ability to quickly penetrate cell membranes and inhibit specific enzymes crucial to cellular respiration. Multiple specific sites of ProClin toxicity prevent microorganisms from developing high levels of resistance, making ProClin highly efficient even over extended storage periods of your IVD components.