Learn More
MSC Earthway Broadcast Spreader, 40 lb, Pneumatic Wheels, Steel T-Bar Handle
Catalog No. 502838041
Encompass
This residential broadcast spreader offers light-duty commercial performance with a rustproof 40 lb (18 kg) poly hopper. It features an EVEN-SPREAD 3-Hole Drop Shut-Off System for uniform application and patented Side Spread Control. With 9 in (22 cm) high-flotation pneumatic tires, it easily navigates various terrains and is suitable for fertilizer, grass seed, salt, grain, and ice melt.
  • 40 lb (18 kg) hopper capacity, rustproof poly construction
  • EVEN-SPREAD 3-hole drop shut-off system for uniform spreading
  • Patented side spread control
  • 100 lb load capacity
  • 9 in (22 cm) high-flotation pneumatic tires on poly rims
  • Epoxy powder-coated chassis
  • Steel "T" bar handle
  • Solid linkage rate control
  • Rate calculation matrix
  • Lifetime technical support
  • 1-year limited warranty
Catalog No. 50-283-8041 Supplier MSC Supplier No. 63821862
This residential broadcast spreader offers light-duty commercial performance with a rustproof 40 lb (18 kg) poly hopper. It features an EVEN-SPREAD 3-Hole Drop Shut-Off System for uniform application and patented Side Spread Control. With 9 in (22 cm) high-flotation pneumatic tires, it easily navigates various terrains and is suitable for fertilizer, grass seed, salt, grain, and ice melt.
  • 40 lb (18 kg) hopper capacity, rustproof poly construction
  • EVEN-SPREAD 3-hole drop shut-off system for uniform spreading
  • Patented side spread control
  • 100 lb load capacity
  • 9 in (22 cm) high-flotation pneumatic tires on poly rims
  • Epoxy powder-coated chassis
  • Steel "T" bar handle
  • Solid linkage rate control
  • Rate calculation matrix
  • Lifetime technical support
  • 1-year limited warranty