Learn More
CPC Scientific H-Thr-Gly-Leu-Leu-Thr-Phe-Leu-Ala-Trp-Trp-His-Glu-Trp-Ala-Ser-Gln-Asp-Ser-Ser-Ser-Thr-Ala-Phe-Glu-Gly-Gly-Thr-Pro-Glu-Leu-Ser-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific CORT005B
Encompass
ONE-LETTER SEQUENCE: TGLLTFLAWWHEWASQDSSSTAFEGGTPELSMOLECULAR FORMULA: C156H219N37O50MOLECULAR WEIGHT:3412.7STORAGE CONDITIONS: -20 5CRESEARCH AREA: NeuropeptidesREFERENCES: L. de Leccea et al., Nature, 381, 242 (1996)
Catalog No. 50-210-9146