Learn More
Alkali Scientific ASI™ Pro-Crystal™ 24 Well Hanging Plate, PS, 18mm, 2μl, 10/Pk, With
SDP
Catalog No. 503028334
Encompass_Preferred
X-rays can be used to analyze the diffraction pattern of atoms in a protein crystal to determine the three-dimensional array of atoms that make up the crystal. The Pro-Crystal ™ plate-24well hanging drop type is the most commonly used form of vapor diffusion method, and it can be easily crystallized and has the advantage of obtaining large and many crystals in a single experiment. • Stackable• Excellent optical properties
Catalog No. 50-302-8334 Supplier Alkali Scientific Supplier No. PCP024S
Add to Cart
Add to Cart
X-rays can be used to analyze the diffraction pattern of atoms in a protein crystal to determine the three-dimensional array of atoms that make up the crystal. The Pro-Crystal ™ plate-24well hanging drop type is the most commonly used form of vapor diffusion method, and it can be easily crystallized and has the advantage of obtaining large and many crystals in a single experiment. • Stackable• Excellent optical properties