Learn More
MSC Liftex Pro-Edge Bridle Web Sling, Type 10, Nylon, 4-Leg, 6 ft., 1 ea.
Catalog No. 5000512500
Encompass
This Type 10 bridle web sling is engineered for heavy-duty lifting applications where load edges contact the sling. Constructed from high-strength nylon, the sling features edge reinforcement with twisted tire cord yarns for enhanced durability. It is designed to meet rigorous safety standards for load lifting and handling.
  • Features 50% better abrasion resistance compared to standard webbing
  • Reinforced with twisted tire cord yarns for edge protection
  • Built with 2 plies for increased load-bearing capacity
  • Includes red core yarn damage indicators for safety monitoring
  • Constructed with durable alloy steel hardware
  • Capacity: 10,300 lbs. at 60°, 8,400 lbs. at 45°, and 6,000 lbs. at 30°
  • Meets OSHA, ASME B30.9, and WSTDA standards
Catalog No. 50-005-12500 Supplier MSC Supplier No. 60015096
This Type 10 bridle web sling is engineered for heavy-duty lifting applications where load edges contact the sling. Constructed from high-strength nylon, the sling features edge reinforcement with twisted tire cord yarns for enhanced durability. It is designed to meet rigorous safety standards for load lifting and handling.
  • Features 50% better abrasion resistance compared to standard webbing
  • Reinforced with twisted tire cord yarns for edge protection
  • Built with 2 plies for increased load-bearing capacity
  • Includes red core yarn damage indicators for safety monitoring
  • Constructed with durable alloy steel hardware
  • Capacity: 10,300 lbs. at 60°, 8,400 lbs. at 45°, and 6,000 lbs. at 30°
  • Meets OSHA, ASME B30.9, and WSTDA standards