Learn More
MSC Twisted Eye & Eye, Type 4 Web Sling: 4' Long, 1" Wide, 3200 lb Vertical Capacity, Nylon
Catalog No. 5000514566
Encompass
This twisted eye and eye type 4 web sling is engineered for secure material handling and lifting applications. Constructed from durable nylon, this two-ply sling offers reliable strength for diverse industrial tasks. It is designed to meet or exceed rigorous OSHA, ASME B30.9, and WSTDA standards for strength and safety.
  • Features a yellow color for easy identification
  • Provides a vertical capacity of 3,200 lb, choker capacity of 2,560 lb, and basket capacity of 6,400 lb
  • Equipped with built-in red core yarn damage indicators for enhanced safety awareness
  • Constructed from durable two-ply nylon material
  • Includes general damage indicators such as holes, tears, cuts, or heat damage
Catalog No. 50-005-14566 Supplier MSC Supplier No. 60007036
This twisted eye and eye type 4 web sling is engineered for secure material handling and lifting applications. Constructed from durable nylon, this two-ply sling offers reliable strength for diverse industrial tasks. It is designed to meet or exceed rigorous OSHA, ASME B30.9, and WSTDA standards for strength and safety.
  • Features a yellow color for easy identification
  • Provides a vertical capacity of 3,200 lb, choker capacity of 2,560 lb, and basket capacity of 6,400 lb
  • Equipped with built-in red core yarn damage indicators for enhanced safety awareness
  • Constructed from durable two-ply nylon material
  • Includes general damage indicators such as holes, tears, cuts, or heat damage