Learn More
MSC Liftex Pro-Edge Polyester Web Sling, 2 Ply, 2 in. x 4 ft., Flat Eyes, 1 ea.
Catalog No. 5000510720
Encompass
This Pro-Edge® polyester web sling is a 2-ply, 2 in. x 4 ft. lifting device designed for superior durability. Constructed with reinforced edge yarns similar to those used in tires, it provides maximum resistance to cutting and abrasion for longer service life compared to standard slings with polymer coatings.
  • Features reinforced edges with twisted tire-cord yarns
  • Provides high resistance to wear, cutting, and abrasion
  • Includes flat eyes on each end
  • Offers a 6,400 lb. vertical capacity
  • Rated for 5,120 lb. choker and 12,800 lb. basket capacities
  • Constructed from 2-ply polyester material
Catalog No. 50-005-10720 Supplier MSC Supplier No. 05068390
This Pro-Edge® polyester web sling is a 2-ply, 2 in. x 4 ft. lifting device designed for superior durability. Constructed with reinforced edge yarns similar to those used in tires, it provides maximum resistance to cutting and abrasion for longer service life compared to standard slings with polymer coatings.
  • Features reinforced edges with twisted tire-cord yarns
  • Provides high resistance to wear, cutting, and abrasion
  • Includes flat eyes on each end
  • Offers a 6,400 lb. vertical capacity
  • Rated for 5,120 lb. choker and 12,800 lb. basket capacities
  • Constructed from 2-ply polyester material