Learn More
MSC PRO HD URINEODOR REMOVER
Catalog No. 502819907
Encompass
This Heavy Duty Urine Stain & Odor Remover utilizes a bio-enzymatic formula to break down organic matter and neutralize odors without harsh chemicals or solvents. Its friendly bacteria produce enzymes that digest organic matter, effectively removing stains and odors from a wide range of surfaces including carpets, upholstery, concrete, vinyl, hardwoods, and tile.
  • Removes pet stains and surface stains on hard and soft surfaces
  • Eliminates odors
  • Features a citrus botanical scent
  • Contains no harsh chemicals
  • Utilizes IP20 technology
  • Made in the USA
Catalog No. 50-281-9907 Supplier MSC Supplier No. 14966303
This Heavy Duty Urine Stain & Odor Remover utilizes a bio-enzymatic formula to break down organic matter and neutralize odors without harsh chemicals or solvents. Its friendly bacteria produce enzymes that digest organic matter, effectively removing stains and odors from a wide range of surfaces including carpets, upholstery, concrete, vinyl, hardwoods, and tile.
  • Removes pet stains and surface stains on hard and soft surfaces
  • Eliminates odors
  • Features a citrus botanical scent
  • Contains no harsh chemicals
  • Utilizes IP20 technology
  • Made in the USA