Learn More
Ted Pella Inc PELCO Pro High Precision, very fine curved tips
SDP
Catalog No. NC0653529
Encompass
Pro High Precision tweezers are made from a non-magnetic, low carbon stainless steel. The tweezers exhibit good corrosion resistance to most chemicals, salts and acids. Typical applications for these tweezers include microscopy preparation, sample handling, precision manufacturing, electronic industry, watch making and general laboratory and medical applications. Although the tweezers are made from a non-magnetic alloy, they are not recommended for use where strong magnetic fields are present. Available in 13 styles and as a set in a top quality plastic wallet with foam lining.
Catalog No. NC0653529 Supplier Ted Pella Inc Supplier No. 53677NM
Pro High Precision tweezers are made from a non-magnetic, low carbon stainless steel. The tweezers exhibit good corrosion resistance to most chemicals, salts and acids. Typical applications for these tweezers include microscopy preparation, sample handling, precision manufacturing, electronic industry, watch making and general laboratory and medical applications. Although the tweezers are made from a non-magnetic alloy, they are not recommended for use where strong magnetic fields are present. Available in 13 styles and as a set in a top quality plastic wallet with foam lining.