Learn More
Meso Scale Discovery V-PLEX Proinflammatory Panel 1 Human Kit, 1 plate
SDP
Catalog No. NC0566622
Encompass
The Proinflammatory Panel 1 (human) Kit provides assay-specific components for the quantitative determination of IFN-gamma, IL-1beta, IL-2, IL-4, IL-6, IL-8, IL-10, IL-12p70, IL-13, and TNF-alpha. This kit may be used for the analysis of all or any combination of these cytokine biomarkers in cell culture supernatants, serum, plasma, and urine.
Catalog No. NC0566622 Supplier Meso Scale Discovery Supplier No. K15049D1
The Proinflammatory Panel 1 (human) Kit provides assay-specific components for the quantitative determination of IFN-gamma, IL-1beta, IL-2, IL-4, IL-6, IL-8, IL-10, IL-12p70, IL-13, and TNF-alpha. This kit may be used for the analysis of all or any combination of these cytokine biomarkers in cell culture supernatants, serum, plasma, and urine.