Learn More
Enzo Life Sciences pro-ISG15 (human), (recombinant) (His-tag) (500 µg)
SDP
Supplier:  Enzo Life Sciences BMLUW92300500
Encompass_Preferred
ISG15 is expressed in its pro-form with a short C-terminal extension, which is processed to expose the C-terminal glycine residue that is essential for conjugation to target proteins. ISG15 is expressed in its pro-form with a short C-terminal extension, which is processed to expose the C-terminal glycine residue that is essential for conjugation to target proteins. Purity: ≥95% (SDS-PAGE). Formulation: Liquid.  In 20mM HEPES, pH 8.0, containing 50mM sodium chloride and 1mM DTT. Source: Produced in E. coli. Human pro-ISG15 is fused at the N-terminus to a His-tag. Long Term Storage: -80°C.
Catalog No. 50-201-2310