Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pro-Lab Diagnostics™ AmnioTest™ Kit Supplies
SDP

Catalog No. 22286159 Shop All Pro-Lab Diagnostics Products
Change view
Click to view available options
pH:
5
5.5
6
6.5
7
7.5
Type:
Color Interpretation Cards
pH 5.0 QC Buffer Solution
pH 5.5 QC Buffer Solution
pH 6.0 QC Buffer Solution
pH 6.5 QC Buffer Solution
pH 7.0 QC Buffer Solution
pH 7.5 QC Buffer Solution
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Type
22-286-159 5.5 pH 5.5 QC Buffer Solution
22-286-158 5 pH 5.0 QC Buffer Solution
22-286-160 6 pH 6.0 QC Buffer Solution
22-286-161 6.5 pH 6.5 QC Buffer Solution
22-286-162 7 pH 7.0 QC Buffer Solution
22-286-164 - Color Interpretation Cards
22-286-163 7.5 pH 7.5 QC Buffer Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 22-286-159 Supplier Pro-Lab Diagnostics™ Supplier No. PL.911
Add to Cart
Add to Cart

Refill buffer solutions and color interpretation cards

Specifications

Type pH 5.5 QC Buffer Solution
For Use With (Application) Use as quality control laboratory reagents to test the performance of AmnioTest™ swabs
pH 5.5
Quantity 2 mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.