Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pro-Lab Diagnostics™ AmnioTest™ Kit Supplies
SDP

Catalog No. 22286164 Shop All Pro-Lab Diagnostics Products
Change view
Click to view available options
Type:
Color Interpretation Cards
pH 5.0 QC Buffer Solution
pH 5.5 QC Buffer Solution
pH 6.0 QC Buffer Solution
pH 6.5 QC Buffer Solution
pH 7.0 QC Buffer Solution
pH 7.5 QC Buffer Solution
pH:
5
5.5
6
6.5
7
7.5
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type pH
22-286-164 Color Interpretation Cards -
22-286-159 pH 5.5 QC Buffer Solution 5.5
22-286-158 pH 5.0 QC Buffer Solution 5
22-286-160 pH 6.0 QC Buffer Solution 6
22-286-161 pH 6.5 QC Buffer Solution 6.5
22-286-162 pH 7.0 QC Buffer Solution 7
22-286-163 pH 7.5 QC Buffer Solution 7.5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 22-286-164 Supplier Pro-Lab Diagnostics™ Supplier No. PL.902
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Refill buffer solutions and color interpretation cards

Specifications

Format Card
Type Color Interpretation Cards
For Use With (Application) Visual comparison of swab color to determine pH level
Quantity 50 Cards
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.