Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pro-Lab Diagnostics™ Copan Microbiological Spreader
SDP

Catalog No. 22029241 Shop All Pro-Lab Diagnostics Products
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-029241 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-029241 Supplier Pro-Lab Diagnostics™ Supplier No. PC174CS05
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-shaped, sterile cell spreader

  • Disposable
  • Hockey stick-shaped
  • Color: Blue
  • Case of 1000

Specifications

Disposable Single-use
Description Individually Wrapped
Quantity 5 cell spreaders per wrapping
Shape L-Shape
Sterility Sterile
Color Blue
Material Polystyrene
Product Type Cell Spreader
For Use With (Application) Spreading microbiological samples on agar surfaces
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.