Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pro-Lab Diagnostics™ Prolex™ - Blue E. coli 0157 Latex Test Reagent Kits
SDP

Catalog No. 22286138 Shop All Pro-Lab Diagnostics Products
Change view
Click to view available options
Quantity:
100 Tests
50 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-286-138 100 Tests
22-286139 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-286-138 Supplier Pro-Lab Diagnostics™ Supplier No. PL.071B
Add to Cart
Add to Cart
This item is not returnable. View return policy

Just one colony is sufficient for the test, enabling even limited growth to be detected

  • Developed to remove the possibility of cross-reactions with E. hermanii and other sorbitol negative Escherichia species
  • Latex agglutination
  • Test results in 2 minutes
  • CLIA Complexity: Non-waived
  • Sensitivity of 100%

Specifications

Content And Storage 2°C to 8°C
Clia Complexity Non-waived
Detectable Analytes E. coli Serogroup O157
Sensitivity 1
Type Blue E. coli O157 Latex Test Reagent Kit
Includes E. coli O157 Latex reagent 6.2mL, negative control Latex reagent 3mL, positive control 3mL, Agglutination cards 50, mixing sticks 150
Quantity 100 Tests
Test Time 2 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.