Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pro-Lab Diagnostics™ Prolex™ Staph Xtra Latex Kits
SDP

Catalog No. 22286204 Shop All Pro-Lab Diagnostics Products
Change view
Click to view available options
Quantity:
100 Tests
300 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-286-204 100 Tests
22-286-202 300 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-286-204 Supplier Pro-Lab Diagnostics™ Supplier No. PL.1080
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Staph Xtra Latex Reagent: 2 x 2.5mL (PL.1080), 2 x 7.5mL (PL.1081)
  • Negative Control Latex Reagent: 5.0mL (PL.1080), 2 x 7.5mL (PL.1081)
  • Agglutination Cards: 20 (PL.1080), 48 (PL.1081)
  • Mixing Sticks: 200 (PL.1080), 300 (PL.1081)

A rapid, sensitive method to distinguish Staphylococcus aureus and methicillin resistant Staphylococcus aureus (MRSA) from other species of Staphylococci

  • Rapid clear results within 20 seconds
  • Uses blue latex particles (CMMP) against a white background

Specifications

Detectable Analytes S. aureus
Type Staph Xtra Latex Kit
Includes 2 x 2.5mL Staph Xtra Latex reagent, 2.5mL negative control Latex reagent, 20 Agglutination cards, 200 mixing sticks
Quantity 100 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.