Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pro-Lab Diagnostics™ Spot Indole Reagent
SDP

Catalog No. 22286249 Shop All Pro-Lab Diagnostics Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-286-249 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-286-249 Supplier Pro-Lab Diagnostics™ Supplier No. PL.39110
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Produces a blue compound upon reaction with indole

  • The detection of indole indicates tryptophan degradation and can be accomplished by the addition of certain aldehydes to form colored end products
  • Spot Indole contains the active ingredient p-dimethylaminocinnamaldehyde (PACA), which reacts with indole to produce a blue compound

Specifications

Content And Storage 2°C to 30°C
Detectable Analytes Indole
Type Spot Indole Reagent
Quantity 10 mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.