| Accession Number |
P41245 |
| Assay Range |
50-18000 pg/mL |
| Assay Sensitivity |
50 pg/mL |
| Calibration Range |
50 pg/mL to 18,000 pg/mL |
| Description |
Pro-MMP-9 ELISA Kit, 96 Assays, 1 Kit, Mouse Species, Streptavidin-HRP Conjugate, Colorimetric Detection Method, Enzyme-Linked Immunosorbent Assay Designed for the Quantitation and Detection of pro-MMP-9 in. Samples Using a Microplate Reader |
| Detectable Analytes |
Pro-MMP-9 |
| Assay |
Solid-phase sandwich ELISA |
| Conjugate |
HRP |
| Detection Method |
Colorimetric |
| Immunoassay Kit Format |
Sandwich ELISA Kit |
| Product Type |
ELISA |
| Protein Family |
Proteases Peptidases & Inhibitors |
| Sample Type |
Plasma, Serum, Supernatant |
| Specificity |
This ELISA kit shows no cross-reactivity with the following cytokines tested: Mouse CD30, L CD30, T CD40, CRG-2, CTACK, CXCL16, Eotaxin, Eotaxin-2, Fas Ligand, Fractalkine, GCSF, GM-CFS, IFN- gamma, IGFBP-3, IGFBP-5, IGFBP-6, IL-1 alpha, IL-1 beta, IL-2 |
| Sufficient For |
5 x 96 assays |
| Target Species |
Mouse |
| For Use With (Application) |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis |
| For Use With (Equipment) |
Absorbance Microplate Reader |
| Gene Alias |
67 kDa matrix metalloproteinase-9; 82 kDa matrix metalloproteinase-9; 92 kDa gelatinase; 92 kDa type IV collagenase; 92kD gelatinase; 92kD type IV collagenase; 92kDa gelatinase; 92kDa type IV collagenase; 92-kDa type IV collagenase; AW743869; B/MMP9; Clg4b; collagenase type IVB; fj05a08; Gel B; gelatinase B; Gelatinase-B; GELB; macrophage gelatinase; MANDP2; matrix metallo protease; matrix metallopeptidase 9; matrix metallopeptidase 9 (gelatinase B, 92kDa gelatinase, 92kDa type IV collagenase); matrix metalloproteinase 9; matrix metalloproteinase 9 (gelatinase B 92-kDa type IV collagenase); matrix metalloproteinase 9 (gelatinase B, 92kDa gelatinase, 92kDa type IV collagenase); matrix metalloproteinase 9 (gelatinase B, 92kDa matrix metalloproteinase 9 (gelatinase B, 92kDa gelatinase, 92kDa type IV collagenase); matrix metalloproteinase 9 (gelatinase B, 92-kDa type IV collagenase); Matrix metalloproteinase9; matrix metalloproteinase-9; MMP; Mmp9; MMP-9; mmp9 protein; MMP-9 protein; MMPs; Preproform of 92-kDa type IV collagenase (MMP-9): N-terminal; pro-MMP-9; type IV collagenase; type V collagenase; wu:fb02g06; wu:fb07b05; wu:fi98c09; wu:fj05a08; ZFMMP-9; zgc:6416567 kDa matrix metalloproteinase-9; 82 kDa matrix metalloproteinase-9; 92 kDa gelatinase; 92 kDa type IV collagenase; 92kD gelatinase; 92kD type IV collagenase; 92kDa gelatinase; 92kDa type IV collagenase; 92-kDa type IV collagenase; AW743869; B/MMP9; Clg4b; collagenase type IVB; fj05a08; Gel B; gelatinase B; Gelatinase-B; GELB; macrophage gelatinase; MANDP2; matrix metallo protease; matrix metallopeptidase 9; matrix metallopeptidase 9 (gelatinase B, 92kDa gelatinase, 92kDa type IV collagenase); matrix metalloproteinase 9; matrix metalloproteinase 9 (gelatinase B 92-kDa type IV collagenase); matrix metalloproteinase 9 (gelatinase B, 92kDa gelatinase, 92kDa type IV collagenase); matrix metalloproteinase 9 (gelatinase B, 92kDa matrix metalloproteinase 9 (gelatinase B, 92kDa gelatinase, 92kDa type IV collagenase); matrix metalloproteinase 9 (gelatinase B, 92-kDa type IV collagenase); Matrix metalloproteinase9; matrix metalloproteinase-9; MMP; Mmp9; MMP-9; mmp9 protein; MMP-9 protein; MMPs; Preproform of 92-kDa type IV collagenase (MMP-9): N-terminal; pro-MMP-9; type IV collagenase; type V collagenase; wu:fb02g06; wu:fb07b05; wu:fi98c09; wu:fj05a08; ZFMMP-9; zgc:64165; AW743869; B/MMP9; Clg4b; Gel B; MMP-9; pro-MMP-9 |
| Gene ID (Entrez) |
17395 |
| Gene Symbol |
MMP9 |
| Host Species |
Mouse |
| Interassay CV |
<12% |
| Intraassay CV |
<10% |
| Kit Contents |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers |
| Label or Dye |
Biotin |
| Protein Aliases |
MMP-9, 92 kDa gelatinase, 92 kDa type IV collagenase, Gelatinase B, GELB |
| Quantity |
5 x 96 Tests |
| Regulatory Status |
RUO |
| Research Discipline |
Bone Research |
| Sample Volume |
Plasma 2 μL, Serum 2 μL, Supernatant 100 μL |
| Shipping Condition |
Wet or Dry Ice |
| Storage Requirements |
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C |
| Synonym |
RP23-61O3.10-001, AW743869, B/MMP9, Clg4b, MMP-9, pro-MMP-9 |
| Target |
MMP-9 |
| Test Time |
1 hr 20 min |
| Total Assay Time |
4 hr 45 min |
|
Show More
Show Less
|
|