Learn More
Sigma Aldrich Fine Chemicals Biosciences Pro PakR distillation pack
Catalog No. 502904990
Encompass
Efficient self-wetting protruded metal dump-packing for distillation columns of 0.75 to 12Ain. diameter. Maximum reflux rate of approximately 3.4AL/h. free space for 0.16Ain.2 size is 94% and for 0.24Ain.2 size is 96%. Each package has a packing volume of approx. 1AL. Packed heights up to 20Aft possible without distortion.
Catalog No. 50-290-4990 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2105521PAK
Efficient self-wetting protruded metal dump-packing for distillation columns of 0.75 to 12Ain. diameter. Maximum reflux rate of approximately 3.4AL/h. free space for 0.16Ain.2 size is 94% and for 0.24Ain.2 size is 96%. Each package has a packing volume of approx. 1AL. Packed heights up to 20Aft possible without distortion.