Learn More
Anatech Ltd Pro-Par Clearant; 1 gallon; Solvents and mounting medium
Catalog No. NC9022529
Encompass_Preferred
Pro-Par Clearant; 1 gal.; Solvents and mounting medium; Unique aliphatic substitute for xylene, Non-irritating, Non-sensitizing, Greaseless and practically odorless; Not derived from citrus oil, Recyclable, Combustible;
Catalog No. NC9022529 Supplier Anatech Ltd Supplier No. 510
Pro-Par Clearant; 1 gal.; Solvents and mounting medium; Unique aliphatic substitute for xylene, Non-irritating, Non-sensitizing, Greaseless and practically odorless; Not derived from citrus oil, Recyclable, Combustible;