Learn More
Anatech Ltd PRO-PAR CLEARANT/4X1GAL
Catalog No. NC9537734
Encompass_Preferred
Unique aliphatic substitute for xylene. Non-irritating, non-sensitizing, greaseless and practically odorless. Not derived from citrus oil. Recyclable. Combustible. Clearant does not remove bound water, and hence keep tissues in a more natural state. Case/4 x 1 gallon
Catalog No. NC9537734 Supplier Anatech Ltd Supplier No. 511
Unique aliphatic substitute for xylene. Non-irritating, non-sensitizing, greaseless and practically odorless. Not derived from citrus oil. Recyclable. Combustible. Clearant does not remove bound water, and hence keep tissues in a more natural state. Case/4 x 1 gallon