Learn More
Medchemexpress LLC Pro-Phe-Arg-AMC acetate | 2237216-30-5 | 99.17% | 635.71 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP26181MG
Encompass_Preferred
Pro-Phe-Arg-AMC acetate is a peptide that serves as a substrate for human glandular kallikrein 2 (hK2). It presents as a solid with a white to off-white appearance. This product is intended strictly for research purposes.
  • Used as a substrate of human glandular kallikrein 2 (hK2)
  • Appears as a solid with a white to off-white color
  • For research use only
Catalog No. 50-004-23469