Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pro-Phe-Arg-AMC acetate | 2237216-30-5 | 99.2% | 635.71 | 5 MG
SDP

Catalog No. 5000422608
Encompass_Preferred

Pro-Phe-Arg-AMC acetate is a peptide that can be used as the substrate of human glandular kallikrein 2 (hK2).

  • Can be used as the substrate of human glandular kallikrein 2 (hK2).
  • Molecular formula: C32H41N7O7.
  • Molecular weight: 635.71.
  • Appears as a white to off-white solid.
  • Soluble in DMSO at 100 mg/mL.
  • Powder storage: -80°C for 2 years, -20°C for 1 year.
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month (sealed storage, away from moisture).

Catalog No. 50-004-22608 Supplier Medchemexpress LLC Supplier No. HYP26185MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pro-Phe-Arg-AMC acetate is a peptide that can be used as the substrate of human glandular kallikrein 2 (hK2).

  • Can be used as the substrate of human glandular kallikrein 2 (hK2).
  • Molecular formula: C32H41N7O7.
  • Molecular weight: 635.71.
  • Appears as a white to off-white solid.
  • Soluble in DMSO at 100 mg/mL.
  • Powder storage: -80°C for 2 years, -20°C for 1 year.
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month (sealed storage, away from moisture).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.