Learn More
Gilson™ Pro Pipette Service for Multichannel Pipettes
Catalog No. SVC101168G Shop All Gilson Products
Change view
Click to view available options
For Use With (Equipment):
12-channel Pipettes
8-channel Pipettes
Multi-channel Pipette
Product Type:
ISO Pipette Pro PM and Calibration Service
Pipette Pro PM and Calibration Service
Service Offering:
After Repair Calibration
As-found/After Repair Calibration
Calibration Points:
2 x 2 Measurements
4 x 2 Measurements
5 x 3 Measurements
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Product Type Service Offering Calibration Points
SVC101168G 12-channel Pipettes ISO Pipette Pro PM and Calibration Service As-found/After Repair Calibration 5 x 3 Measurements
SVC101156G 12-channel Pipettes Pipette Pro PM and Calibration Service As-found/After Repair Calibration 5 x 3 Measurements
SVC101144G 12-channel Pipettes ISO Pipette Pro PM and Calibration Service As-found/After Repair Calibration 4 x 2 Measurements
SVC101132G 12-channel Pipettes Pipette Pro PM and Calibration Service As-found/After Repair Calibration 4 x 2 Measurements
SVC101108G 12-channel Pipettes Pipette Pro PM and Calibration Service After Repair Calibration 2 x 2 Measurements
SVC101120G 12-channel Pipettes Pipette Pro PM and Calibration Service After Repair Calibration 4 x 2 Measurements
SVC101167G 8-channel Pipettes ISO Pipette Pro PM and Calibration Service As-found/After Repair Calibration 5 x 3 Measurements
SVC101155G 8-channel Pipettes Pipette Pro PM and Calibration Service As-found/After Repair Calibration 5 x 3 Measurements
SVC101143G 8-channel Pipettes ISO Pipette Pro PM and Calibration Service As-found/After Repair Calibration 4 x 2 Measurements
SVC101131G 8-channel Pipettes Pipette Pro PM and Calibration Service As-found/After Repair Calibration 4 x 2 Measurements
SVC101107G 8-channel Pipettes Pipette Pro PM and Calibration Service After Repair Calibration 2 x 2 Measurements
SVC101119G 8-channel Pipettes Pipette Pro PM and Calibration Service After Repair Calibration 4 x 2 Measurements
SVC101164G Multi-channel Pipette ISO Pipette Pro PM and Calibration Service As-found/After Repair Calibration 5 x 3 Measurements
SVC101152G Multi-channel Pipette Pipette Pro PM and Calibration Service As-found/After Repair Calibration 5 x 3 Measurements
SVC101140G Multi-channel Pipette ISO Pipette Pro PM and Calibration Service As-found/After Repair Calibration 4 x 2 Measurements
SVC101128G Multi-channel Pipette Pipette Pro PM and Calibration Service As-found/After Repair Calibration 4 x 2 Measurements
SVC101104G Multi-channel Pipette Pipette Pro PM and Calibration Service After Repair Calibration 2 x 2 Measurements
SVC101116G Multi-channel Pipette Pipette Pro PM and Calibration Service After Repair Calibration 4 x 2 Measurements
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Catalog No. SVC101168G Supplier Gilson™ Supplier No. SVC101168
Add to Cart
Add to Cart
Gilson Pro Pipette Service offers quality service, preventative maintenance, and calibration to ensure pipettes lasts for years to come.
  • Visual and Functional Inspection
  • Internal and External Cleaning
  • Leak Test
  • Gravimetric Check
  • Preventative Maintenance
  • Re-grease as needed
  • Pipette Label
  • Mail-In or Onsite Service Available
  • Calibration and Detailed Certificate
  • ISO 17025 Calibration Available
  • 5 day turn around time
Specifications
For Use With (Equipment) 12-channel Pipettes
Certifications/Compliance ISO 17025 Accredited Service
Description Data collected before and after repair
Calibration Points 5 x 3 Measurements
Duration 5 Days
Product Type ISO Pipette Pro PM and Calibration Service
Service Offering As-found/After Repair Calibration