Learn More
Gilson™ Pro Pipette Service for Single Channel Pipettes
Catalog No. SVC101151G Shop All Gilson Products
Change view
Click to view available options
Product Type:
ISO Preventive Maintenance and Calibration
Preventive Maintenance and Calibration
Service Offering:
After Repair Calibration
As-found/After Repair Calibration
Calibration Points:
2 x 2 Measurements
4 x 2 Measurements
5 x 3 Measurements
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Service Offering Calibration Points
SVC101151G Preventive Maintenance and Calibration As-found/After Repair Calibration 5 x 3 Measurements
SVC101163G ISO Preventive Maintenance and Calibration As-found/After Repair Calibration 5 x 3 Measurements
SVC101139G ISO Preventive Maintenance and Calibration As-found/After Repair Calibration 4 x 2 Measurements
SVC101127G Preventive Maintenance and Calibration As-found/After Repair Calibration 4 x 2 Measurements
SVC101103G Preventive Maintenance and Calibration After Repair Calibration 2 x 2 Measurements
SVC101115G Preventive Maintenance and Calibration After Repair Calibration 4 x 2 Measurements
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. SVC101151G Supplier Gilson™ Supplier No. SVC101151
Add to Cart
Add to Cart
Gilson Pro Pipette Service offers quality service, preventative maintenance, and calibration to ensure pipettes lasts for years to come.
  • Visual and Functional Inspection
  • Internal and External Cleaning
  • Leak Test
  • Gravimetric Check
  • Preventative Maintenance
  • Re-grease as needed
  • Pipette Label
  • Mail-In or Onsite Service Available
  • Calibration and Detailed Certificate
  • ISO 17025 Calibration Available
  • 5 day turn around time
Specifications
For Use With (Equipment) Single-channel Pipette
Certifications/Compliance ISO 17025 Accredited Service
Description Data collected before and after repair
Calibration Points 5 x 3 Measurements
Duration 5 Days
Calibration Type Gravimetric
Product Type Preventive Maintenance and Calibration
Product Line Pipette Pro
Service Offering As-found/After Repair Calibration