Learn More
HANNA INSTRUMENTS INC Professional Portable Meat pH Mete
SDP
Catalog No. NC1632754
Encompass
The HI98163 is a rugged, waterproof, Foodcare portable pH meter that measures pH and temperature using the specialized FC2323 meat pH electrode with a stainless steel piercing blade. This professional, waterproof meter complies with IP67 standards. The HI98163 is supplied with all necessary accessories to perform a pH/temperature measurement packaged into a durable thermoformed carrying case that holds the meters, probes and calibration buffers securely in place.Quick Connect DIN ConnectorUSB Connection for Data TransferDedicated Help Button
Catalog No. NC1632754 Supplier HANNA INSTRUMENTS INC Supplier No. HI98163
The HI98163 is a rugged, waterproof, Foodcare portable pH meter that measures pH and temperature using the specialized FC2323 meat pH electrode with a stainless steel piercing blade. This professional, waterproof meter complies with IP67 standards. The HI98163 is supplied with all necessary accessories to perform a pH/temperature measurement packaged into a durable thermoformed carrying case that holds the meters, probes and calibration buffers securely in place.Quick Connect DIN ConnectorUSB Connection for Data TransferDedicated Help Button