Learn More
MSC PRO-SAFE Aluminized Pants, Aluminized Blended Kevlar, Large, 1 ea
Catalog No. 5000687653
Encompass
This product is a pair of aluminized pants made from an aluminized blended Kevlar material, designed for a waist size of 36 to 38 in.
  • Large size
  • Designed for 36 to 38 in. waist
  • Made from aluminized blended Kevlar
  • Silver color
  • Material weight of 19.0 oz
  • Manufactured in the United States
Catalog No. 50-006-87653 Supplier MSC Supplier No. 54165253
This product is a pair of aluminized pants made from an aluminized blended Kevlar material, designed for a waist size of 36 to 38 in.
  • Large size
  • Designed for 36 to 38 in. waist
  • Made from aluminized blended Kevlar
  • Silver color
  • Material weight of 19.0 oz
  • Manufactured in the United States