Learn More
MSC PRO-SAFE Aluminized Pants, Aluminized Carbon Kevlar, 36 in., 1 ea.
Catalog No. 5000686107
Encompass
These aluminized pants offer superior protection, constructed from Aluminized Carbon Kevlar for high heat resistance in demanding industrial environments.
  • Garment type: aluminized
  • Colors: silver, yellow, black
  • Length: 36 in.
  • Package quantity: 1
  • Country of origin: United States
Catalog No. 50-006-86107 Supplier MSC Supplier No. 54165360
These aluminized pants offer superior protection, constructed from Aluminized Carbon Kevlar for high heat resistance in demanding industrial environments.
  • Garment type: aluminized
  • Colors: silver, yellow, black
  • Length: 36 in.
  • Package quantity: 1
  • Country of origin: United States