Learn More
MSC PRO-SAFE Aluminized Pants, Medium, Aluminized Carbon Kevlar, 1 Pair
Catalog No. 5000673741
Encompass
PRO-SAFE aluminized pants are designed for protection in environments requiring high heat resistance. Constructed from advanced aluminized carbon Kevlar, these pants offer superior thermal protection for workers in hazardous conditions.
  • Size: Medium
  • Material: Aluminized carbon Kevlar
  • Color: Silver
  • Material weight: 19.0 oz
  • Country of origin: United States
Catalog No. 50-006-73741 Supplier MSC Supplier No. 54165220
PRO-SAFE aluminized pants are designed for protection in environments requiring high heat resistance. Constructed from advanced aluminized carbon Kevlar, these pants offer superior thermal protection for workers in hazardous conditions.
  • Size: Medium
  • Material: Aluminized carbon Kevlar
  • Color: Silver
  • Material weight: 19.0 oz
  • Country of origin: United States