Learn More
MSC PRO-SAFE Anti-Fatigue Mat, 12 ft, 3 ft, 1 ea
Catalog No. 5000792781
Encompass
PRO-SAFE anti-fatigue mats are engineered for comfort and safety in demanding dry work environments. Constructed with a composite vinyl material, these heavy-duty mats feature a diamond plate surface and beveled edges to enhance user stability and prevent tripping hazards.
  • Designed for heavy-duty use in dry work areas
  • Features a durable vinyl surface and vinyl sponge base for composite construction
  • Beveled edges on all four sides prevent tripping
  • Diamond plate surface pattern
  • Includes two colored borders (black and yellow)
  • Made from recycled content
Catalog No. 50-007-92781 Supplier MSC Supplier No. 88267166
PRO-SAFE anti-fatigue mats are engineered for comfort and safety in demanding dry work environments. Constructed with a composite vinyl material, these heavy-duty mats feature a diamond plate surface and beveled edges to enhance user stability and prevent tripping hazards.
  • Designed for heavy-duty use in dry work areas
  • Features a durable vinyl surface and vinyl sponge base for composite construction
  • Beveled edges on all four sides prevent tripping
  • Diamond plate surface pattern
  • Includes two colored borders (black and yellow)
  • Made from recycled content